CH4375444698 ChemBridge Corporation

3-(4-methoxy-3-methylphenyl)acrylic acid

  • MDL number MFCD03002824
  • Cas Number : 175675-62-4
  • Molecular Weight : 192.2112

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6072393

Safety Information

MSDS https://www.hit2lead.com/sds/6072393_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart