CH4375444706 ChemBridge Corporation

6-chloro-1-isoindolinone

  • MDL number MFCD08234734
  • Cas Number : 58083-59-3
  • Molecular Weight : 167.5923

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4046843

Safety Information

MSDS https://www.hit2lead.com/sds/4046843_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart