CH4375444714 ChemBridge Corporation

2-bromo-4-methylquinoline

  • MDL number MFCD08705663
  • Cas Number : 64658-04-4
  • Molecular Weight : 222.0812

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4049766

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4049766
Availability details for
Quantity
  • Availability
  • Add to Cart