CH4388365713 ChemBridge Corporation

(3S)-1-methyl-3-phenylpiperazine

  • MDL number MFCD09055250
  • Cas Number : 931115-08-1
  • Molecular Weight : 176.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4004801

Safety Information

MSDS https://www.hit2lead.com/sds/4004801_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart