CH4388365722 ChemBridge Corporation

2-piperazin-1-ylcyclohexanol

  • MDL number MFCD07801128
  • Cas Number : 759402-42-1
  • Molecular Weight : 184.2786

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4010894

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4010894
Availability details for
Quantity
  • Availability
  • Add to Cart