CH4388365724 ChemBridge Corporation

4,7,7-trimethyl-3-oxobicyclo[2.2.1]heptane-1-carboxylic acid

  • MDL number MFCD08752991
  • Cas Number : 6703-31-7
  • Molecular Weight : 196.2429

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4010937

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4010937
Availability details for
Quantity
  • Availability
  • Add to Cart