CH4388365726 ChemBridge Corporation

2-(aminosulfonyl)benzoic acid

  • MDL number MFCD00127704
  • Cas Number : 632-24-6
  • Molecular Weight : 201.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4010962

Safety Information

MSDS https://www.hit2lead.com/sds/4010962_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart