CH4388365738 ChemBridge Corporation

1-isopropyl-1H-1,2,3-triazol-4-amine

  • MDL number MFCD09864367
  • Cas Number : 959237-92-4
  • Molecular Weight : 126.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4018384

Safety Information

MSDS https://www.hit2lead.com/sds/4018384_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart