CH4388365742 ChemBridge Corporation

5-bromoquinolin-8-ol

  • MDL number MFCD00457011
  • Cas Number : 1198-14-7
  • Molecular Weight : 224.054

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4023291

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4023291
Availability details for
Quantity
  • Availability
  • Add to Cart