CH4388365750 ChemBridge Corporation

5-bromo-8-quinolinamine

  • MDL number MFCD01571832
  • Cas Number : 53472-18-7
  • Molecular Weight : 223.0693

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4031169

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4031169
Availability details for
Quantity
  • Availability
  • Add to Cart