CH4388365770 ChemBridge Corporation

6-bromo-2-methyl[1,3]thiazolo[5,4-b]pyridine

  • MDL number MFCD07375016
  • Cas Number : 886372-92-5
  • Molecular Weight : 229.097

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4047192

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4047192
Availability details for
Quantity
  • Availability
  • Add to Cart