CH4388365780 ChemBridge Corporation

5-bromo-4-methoxy-6-methylpyrimidine

  • MDL number MFCD19382397
  • Cas Number : 4319-87-3
  • Molecular Weight : 203.0366

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4048808

Safety Information

MSDS https://www.hit2lead.com/sds/4048808_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart