CH4388365782 ChemBridge Corporation

cis-3-(methoxycarbonyl)cyclopentanecarboxylic acid

  • MDL number MFCD01311176
  • Cas Number : 96382-85-3
  • Molecular Weight : 172.1785

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4049737

Safety Information

MSDS https://www.hit2lead.com/sds/4049737_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart