CH4388365785 ChemBridge Corporation

3,6-dichloro-2-methylpyridine

  • MDL number MFCD00778766
  • Cas Number : 123280-64-8
  • Molecular Weight : 162.0166

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4050886

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4050886
Availability details for
Quantity
  • Availability
  • Add to Cart