CH4388365786 ChemBridge Corporation

5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)-2,3-dihydro-1,4-dioxine

  • MDL number MFCD13182121
  • Cas Number : 1046811-97-5
  • Molecular Weight : 212.05

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4052968

Safety Information

MSDS https://www.hit2lead.com/sds/4052968_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart