CH4388365804 ChemBridge Corporation

(methylamino)(pyridin-3-yl)acetic acid

  • MDL number MFCD05864551
  • Cas Number : 115200-98-1
  • Molecular Weight : 166.17

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4141573

Safety Information

MSDS https://www.hit2lead.com/sds/4141573_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart