CH4388365806 ChemBridge Corporation

(3-phenylisoxazol-5-yl)boronic acid

  • MDL number MFCD04039036
  • Cas Number : 5868-54-2
  • Molecular Weight : 188.97

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4201613

Safety Information

MSDS https://www.hit2lead.com/sds/4201613_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart