CH4388365809 ChemBridge Corporation

5-(methoxymethyl)-2-furaldehyde

  • MDL number MFCD02253954
  • Cas Number : 1917-64-2
  • Molecular Weight : 140.13

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4301975

Safety Information

MSDS https://www.hit2lead.com/sds/4301975_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart