CH4388365818 ChemBridge Corporation

(1-methyl-1H-pyrrol-3-yl)acetic acid

  • MDL number MFCD08060084
  • Cas Number : 915919-88-9
  • Molecular Weight : 139.15

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4400835

Safety Information

MSDS https://www.hit2lead.com/sds/4400835_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart