CH4388365819 ChemBridge Corporation

2-phenyl-1H-imidazole-4-carboxylic acid

  • MDL number MFCD06203404
  • Cas Number : 77498-98-7
  • Molecular Weight : 188.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4401605

Safety Information

MSDS https://www.hit2lead.com/sds/4401605_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart