CH4388365828 ChemBridge Corporation

4-hydroxy-2H-thiochromen-2-one

  • MDL number MFCD00463527
  • Cas Number : 16854-67-4
  • Molecular Weight : 178.20

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5102960

Safety Information

MSDS https://www.hit2lead.com/sds/5102960_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart