CH4388365835 ChemBridge Corporation

1,3-dimethyl-2-phenylpiperidin-4-one

  • MDL number MFCD00543850
  • Cas Number : 161894-21-9
  • Molecular Weight : 203.28

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5118842

Safety Information

MSDS https://www.hit2lead.com/sds/5118842_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart