CH4388365836 ChemBridge Corporation

4-hydroxy-4,4-diphenylbut-2-ynoic acid

  • MDL number MFCD00810269
  • Cas Number : 29262-25-7
  • Molecular Weight : 252.26

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5118935

Safety Information

MSDS https://www.hit2lead.com/sds/5118935_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart