CH4388365841 ChemBridge Corporation

1-ethyl-2-methyl-1H-benzimidazol-5-ol

  • MDL number MFCD00159951
  • Cas Number : 82612-27-9
  • Molecular Weight : 176.21

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5157009

Safety Information

MSDS https://www.hit2lead.com/sds/5157009_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart