CH4388365844 ChemBridge Corporation

2-(4-chlorophenoxy)-3-(dimethylamino)propanoic acid

  • MDL number MFCD00559639
  • Cas Number : 2130-72-5
  • Molecular Weight : 243.68

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5213112

Safety Information

MSDS https://www.hit2lead.com/sds/5213112_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart