CH4388365849 ChemBridge Corporation

1-(3-butoxyphenyl)cyclopentanecarboxylic acid

  • MDL number MFCD00431948
  • Cas Number : 74316-94-2
  • Molecular Weight : 262.34

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5238076

Safety Information

MSDS https://www.hit2lead.com/sds/5238076_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart