CH4388365856 ChemBridge Corporation

5-(morpholin-4-ylcarbonyl)adamantan-2-one

  • MDL number MFCD03105593
  • Cas Number : 174972-27-1
  • Molecular Weight : 263.33

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5357760

Safety Information

MSDS https://www.hit2lead.com/sds/5357760_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart