CH4388365859 ChemBridge Corporation

5-(acetyloxy)nicotinic acid

  • MDL number MFCD01532498
  • Cas Number : 325970-19-2
  • Molecular Weight : 181.14

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5511863

Safety Information

MSDS https://www.hit2lead.com/sds/5511863_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart