CH4388365866 ChemBridge Corporation

2,5-dioxo-1-phenyl-1,2,5,6,7,8-hexahydroquinoline-3-carboxylic acid

  • MDL number MFCD00981288
  • Cas Number : 125885-51-0
  • Molecular Weight : 283.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5561748

Safety Information

MSDS https://www.hit2lead.com/sds/5561748_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart