CH4388365870 ChemBridge Corporation

9H-carbazol-3-ylacetic acid

  • MDL number MFCD01469717
  • Cas Number : 374916-11-7
  • Molecular Weight : 225.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5584651

Safety Information

MSDS https://www.hit2lead.com/sds/5584651_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart