CH4388365878 ChemBridge Corporation

4-(3-bromo-4-methoxyphenyl)-1,3-thiazol-2-amine

  • MDL number MFCD01539893
  • Cas Number : 189011-00-5
  • Molecular Weight : 285.16

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5846891

Safety Information

MSDS https://www.hit2lead.com/sds/5846891_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart