CH4388365885 ChemBridge Corporation

2-[(2-hydroxyethyl)sulfinyl]benzoic acid

  • MDL number MFCD00764798
  • Cas Number : 325703-91-1
  • Molecular Weight : 214.23

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/5916945

Safety Information

MSDS https://www.hit2lead.com/sds/5916945_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart