CH4388365889 ChemBridge Corporation

1-ethyl-6-(2-furyl)-4-oxo-1,4-dihydroquinoline-3-carboxylic acid

  • MDL number MFCD02053223
  • Cas Number : 102269-27-2
  • Molecular Weight : 283.27

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6044230

Safety Information

MSDS https://www.hit2lead.com/sds/6044230_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart