CH4388365896 ChemBridge Corporation

3-(5,7-dioxo-5,7-dihydro-6H-pyrrolo[3,4-b]pyridin-6-yl)benzoic acid

  • MDL number MFCD02031683
  • Cas Number : 294892-44-7
  • Molecular Weight : 268.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6096141

Safety Information

MSDS https://www.hit2lead.com/sds/6096141_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart