CH4388365897 ChemBridge Corporation

3-(5-formyl-2-furyl)benzenesulfonamide

  • MDL number MFCD02063358
  • Cas Number : 306935-94-4
  • Molecular Weight : 251.25

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6102433

Safety Information

MSDS https://www.hit2lead.com/sds/6102433_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart