CH4388365900 ChemBridge Corporation

6-methyl-4-quinazolinamine

  • MDL number MFCD02343058
  • Cas Number : 21419-47-6
  • Molecular Weight : 159.18

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6142951

Safety Information

MSDS https://www.hit2lead.com/sds/6142951_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart