CH4388365906 ChemBridge Corporation

3-(1H-benzimidazol-2-yl)benzoic acid

  • MDL number MFCD02323224
  • Cas Number : 402944-81-4
  • Molecular Weight : 238.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6435212

Safety Information

MSDS https://www.hit2lead.com/sds/6435212_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart