CH4388365910 ChemBridge Corporation

rac-(2S,4aR,8aR)-2-methyloctahydro-4(1H)-quinolinone

  • MDL number MFCD10686640
  • Cas Number : 1932006-19-3
  • Molecular Weight : 167.24

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6482384

Safety Information

MSDS https://www.hit2lead.com/sds/6482384_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart