CH4388365915 ChemBridge Corporation

4-(2,4,6-trioxotetrahydropyrimidin-1(2H)-yl)benzoic acid

  • MDL number MFCD00845866
  • Cas Number : 54185-33-0
  • Molecular Weight : 248.19

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6630114

Safety Information

MSDS https://www.hit2lead.com/sds/6630114_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart