CH4388365916 ChemBridge Corporation

N-2-biphenylyl-2-chloroacetamide

  • MDL number MFCD00859219
  • Cas Number : 23088-28-0
  • Molecular Weight : 245.70

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6636578

Safety Information

MSDS https://www.hit2lead.com/sds/6636578_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart