CH4388365918 ChemBridge Corporation

1-(4-isopropoxyphenyl)cyclopentanecarboxylic acid

  • MDL number MFCD03161239
  • Cas Number : 74316-96-4
  • Molecular Weight : 248.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6666041

Safety Information

MSDS https://www.hit2lead.com/sds/6666041_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart