CH4388365924 ChemBridge Corporation

2-(4-methylpiperazin-1-yl)-4-oxo-4H-pyrido[1,2-a]pyrimidine-3-carbaldehyde

  • MDL number MFCD03285279
  • Cas Number : 154816-49-6
  • Molecular Weight : 272.30

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/6895554

Safety Information

MSDS https://www.hit2lead.com/sds/6895554_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart