CH4388365939 ChemBridge Corporation

3-bromo-5,7-dimethylpyrazolo[1,5-a]pyrimidin-2(1H)-one

  • MDL number MFCD00501929
  • Cas Number : 66383-54-8
  • Molecular Weight : 242.07

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/7978855

Safety Information

MSDS https://www.hit2lead.com/sds/7978855_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart