CH4388365941 ChemBridge Corporation

methyl 2-amino-3-bromo-5-methylbenzoate

  • MDL number MFCD07439821
  • Cas Number : 206551-32-8
  • Molecular Weight : 244.08

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 0.95
Salt Data FREE
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/9033713

Safety Information

MSDS https://www.hit2lead.com/sds/9033713_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart