CH4394346199 ChemBridge Corporation

3-(difluoromethyl)-1-methyl-1H-pyrazole-5-carboxylic acid

  • MDL number MFCD09971822
  • Cas Number : 1052617-38-5
  • Molecular Weight : 176.1208

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4039760

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4039760
Availability details for
Quantity
  • Availability
  • Add to Cart