CH4394346216 ChemBridge Corporation

4-(methylthio)-2(1H)-pyrimidinone

  • MDL number MFCD07732327
  • Cas Number : 35551-31-6
  • Molecular Weight : 142.1789

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4048071

Safety Information

MSDS https://www.hit2lead.com/sds/4048071_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart