CH4394346220 ChemBridge Corporation

(6-fluoro-8-quinolinyl)boronic acid

  • MDL number MFCD11504849
  • Cas Number : 1072951-44-0
  • Molecular Weight : 190.9668

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4048990

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4048990
Availability details for
Quantity
  • Availability
  • Add to Cart