CH4394346221 ChemBridge Corporation

2-bromo-1-phenylethanol

  • MDL number MFCD00540454
  • Cas Number : 2425-28-7
  • Molecular Weight : 201.0605

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. -20C
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4049086

Safety Information

MSDS https://www.hit2lead.com/sds/4049086_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart