CH4394346224 ChemBridge Corporation

3-amino-1-methyl-1H-pyrazole-4-carboxylic acid

  • MDL number MFCD14519628
  • Cas Number : 151733-97-0
  • Molecular Weight : 141.128

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. Room Temperature
Shipping Temperature Room Temperature
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4049817

Safety Information

MSDS https://www.hit2lead.com/building-blocks/4049817
Availability details for
Quantity
  • Availability
  • Add to Cart