CH4407083220 ChemBridge Corporation

tert-butyl 2-(5-bromo-2-pyrimidinyl)hydrazinecarboxylate

  • MDL number MFCD26130617
  • Cas Number : 1452182-34-1
  • Molecular Weight : 289.1291

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data FREE
Storage temp. RT
Physical Form solid
Shipping Temperature RT
Country of Origin Russia
Product Link https://www.hit2lead.com/building-blocks/4085421

Safety Information

MSDS https://www.hit2lead.com/sds/4085421_sds.pdf
Availability details for
Quantity
  • Availability
  • Add to Cart